Learn More
Invitrogen™ ACTRT2 Polyclonal Antibody

Description
Immunogen sequence: NGSGFCKAGL SGEFGPRHMV SSIVGHLKFQ APSAEANQKK YFVGEEALYK QEALQLHSPF ERGLITGWDD VERLWKHLFE WELGVKPSDQ PLLATEPSLN PRENREKMAE VMFENFGVPA FYLSDQ Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
Specifications
Specifications
| Antigen | ACTRT2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ACTRT2 |
| Gene Accession No. | Q8TDY3 |
| Gene Alias | 1700052K15Rik; actin related protein M2 (Arpm2); actin related protein T2; actin-related protein hArpM2; actin-related protein M2; Actin-related protein T2; Actrt2; ARPM2; ARPT2; ARP-T2; HARPM2 |
| Gene Symbols | ACTRT2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.