Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACTRT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555228 Supplier Invitrogen™ Supplier No. PA555228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALEPEKELSR RPEEVLREYK LPDGNIISLG DPLHQAPEAL FVPQQLGSQS PGLSNMVSSS ITKCDTDIQK ILFGEIVLSG GTTLFHGLDD RLLKELEQLA SKDTPIKITA P Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.

The protein encoded by this intronless gene belongs to the actin family. Studies have shown that this protein may be involved in cytoskeletal organization similar to other cytoplasmic actin-related protein (ARP) subfamily members. Antibody raised against the human protein has been used to detect the protein by immunoblotting and immunofluorescence microscopy, demonstrating its specific synthesis in the testis, late in spermatid differentiation, and its localization in the calyx.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACTRT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACTRT2
Gene Accession No. Q8TDY3
Gene Alias 1700052K15Rik; actin related protein M2 (Arpm2); actin related protein T2; actin-related protein hArpM2; actin-related protein M2; Actin-related protein T2; Actrt2; ARPM2; ARPT2; ARP-T2; HARPM2
Gene Symbols ACTRT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACTRT2. Recombinant protein control fragment (Product #RP-88853).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140625
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.