Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACTRT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557411
Description
Immunogen sequence: QGGLELCVGD QAQDWRSSLF ISYPVERGLI TSWEDMEIMW KHIYDYNLKL KPCDGPVLIT EPALNPLANR QQITEMF Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.
ACTRT3 is a protein coding gene.
Specifications
ACTRT3 | |
Polyclonal | |
Unconjugated | |
ACTRT3 | |
1700119I24Rik; actin related protein M1; actin related protein T3; actin, gamma 1 propeptide; actin-related protein M1; actin-related protein M1 homolog; actin-related protein T3; ACTRT3; ARPM1; ARP-T3; RGD1561457 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84517 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYD9 | |
ACTRT3 | |
Recombinant protein corresponding to Human ACTRT3. Recombinant protein control fragment (Product #RP-96470). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction