Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518842
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518842 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518842 Supplier Invitrogen™ Supplier No. PA518842
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors. The ACVR1 gene encodes activin A type I receptor, which signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ACVR1
Gene Accession No. Q04771
Gene Alias activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7L; TSRI; TSR-I; type I TGF B receptor
Gene Symbols ACVR1
Host Species Goat
Immunogen Synthetic peptide sequence (RKFKRRNQERLNPRD) corresponding to the internal amino acids of ACVR1 (aa 147-162).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.