Learn More
Invitrogen™ ACVR1 Polyclonal Antibody

Description
A suggested positive control is A549 cell lysate. PA5-20604 can be used with blocking peptide PEP-0724.
Specifications
Specifications
| Antigen | ACVR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ACVR1 |
| Gene Accession No. | P37172, Q04771 |
| Gene Alias | activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7L; TSRI; TSR-I; type I TGF B receptor |
| Gene Symbols | ACVR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.