Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACVR1B Monoclonal Antibody (179CT11.2.1)
GREENER_CHOICE

Catalog No. PIMA537487
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537487 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537487 Supplier Thermo Scientific Supplier No. MA537487
Add to Cart
Add to Cart

ACVR1B Monoclonal Antibody (179CT11.2.1)

ACVR1B Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR1B
Applications Western Blot
Classification Monoclonal
Clone 179CT11.2.1
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ACVR1B
Gene Accession No. Q61271
Gene Alias 6820432J04; activin A receptor type 1B; activin A receptor type IB; activin A receptor, type 1B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; activin receptor type IB; Activin receptor type-1B; activin receptor-like kinase 4; ACTRI; ActRIB; ActR-IB; ACVR1A; Acvr1b; ACVRLK2; ACVRLK4; ALK2; Alk4; ALK-4; FOP; Serine/threonine-protein kinase receptor R2; SKR1; SKR2; TSRI
Gene Symbols ACVR1B
Host Species Mouse
Immunogen ACVR1B recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11479
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.