Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVR1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101107
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101107 Supplier Invitrogen™ Supplier No. PA5101107
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACV1B.

ACVR1B is a serine/threonine kinase involved in signal transduction and cellular communication. Several truncated ACVR1B isoforms have been reported to be exclusively expressed in human pituitary tumors. Mutations of ACVR1B have been observed in pancreatic adenocarcinomas. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. ACVR1B encodes activin A type IB receptor, composed of 11 exons. Alternative splicing and alternative polyadenylation result in 3 fully described transcript variants. The mRNA expression of variants 1, 2, and 3 is confirmed, and a potential fourth variant contains an alternative exon 8 and lacks exons 9 through 11, but its mRNA expression has not been confirmed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR1B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACVR1B
Gene Accession No. P36896, P80202, Q61271
Gene Alias 6820432J04; activin A receptor type 1B; activin A receptor type IB; activin A receptor, type 1B; activin A receptor, type IB; activin A receptor, type II-like kinase 4; activin receptor type IB; Activin receptor type-1B; activin receptor-like kinase 4; ACTRI; ActRIB; ActR-IB; ACVR1A; Acvr1b; ACVRLK2; ACVRLK4; ALK2; Alk4; ALK-4; FOP; Serine/threonine-protein kinase receptor R2; SKR1; SKR2; TSRI
Gene Symbols ACVR1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACVR1B(Accession P36896), corresponding to amino acid residues R91-I141.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11479, 29381, 91
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.