Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVR1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102319 Supplier Invitrogen™ Supplier No. PA5102319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACVR1C.

ACVR1C, a serine/threonine protein kinase, is a type I receptor for the TGFB family of signaling molecules. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors. ACVR1C is a receptor for activin AB, activin B and NODAL. This protein plays a role in cell differentiation, growth arrest and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR1C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACVR1C
Gene Accession No. P70539, Q8K348, Q8NER5
Gene Alias activin A receptor type 1C; activin A receptor type IC; activin A receptor, type IC; activin receptor type IC; Activin receptor type-1C; activin receptor-like kinase 7; ACTRI; ACTR-IC; ACVR1A; acvr1c; acvr1c {ECO:0000312; ACVRLK2; ACVRLK7; ALK2; Alk7; ALK-7; C230097P10; EMBL:AAC52803.1}; FOP; habrec1; SKR1; TGF-beta type 1 receptor; TSRI
Gene Symbols ACVR1C
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACVR1C(Accession Q8NER5), corresponding to amino acid residues P417-A467.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130399, 245921, 269275
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.