Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACVR1C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587475
Description
Immunogen sequence: LSPGLKCVCL LCDSSNFTCQ TEGACWASVM LTNGKEQVIK SCVSLPELNA QVFCHSSNNV TKTECCFTDF CNNITLHLPT ASPNAPKLGP ME; Positive Samples: SH-SY5Y; Cellular Location: Membrane, Single-pass type I membrane protein.
ACVR1C, a serine/threonine protein kinase, is a type I receptor for the TGFB family of signaling molecules. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3, which then translocate to the nucleus and interact directly with DNA or in complex with other transcription factors. ACVR1C is a receptor for activin AB, activin B and NODAL. This protein plays a role in cell differentiation, growth arrest and apoptosis.
Specifications
ACVR1C | |
Polyclonal | |
Unconjugated | |
ACVR1C | |
activin A receptor type 1C; activin A receptor type IC; activin A receptor, type IC; activin receptor type IC; Activin receptor type-1C; activin receptor-like kinase 7; ACTRI; ACTR-IC; ACVR1A; acvr1c; acvr1c {ECO:0000312; ACVRLK2; ACVRLK7; ALK2; Alk7; ALK-7; C230097P10; EMBL:AAC52803.1}; FOP; habrec1; SKR1; TGF-beta type 1 receptor; TSRI | |
Rabbit | |
Affinity Chromatography | |
RUO | |
130399, 245921, 269275 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.17 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P70539, Q8K348, Q8NER5 | |
ACVR1C | |
Recombinant fusion protein containing a sequence corresponding to amino acids 22-113 of human ACVR1C (NP_6603022). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction