Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVR2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114823 Supplier Invitrogen™ Supplier No. PA5114823
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACVR2A.

Activin A receptor kinase (ACVR2A) is a type II member of the TGF-beta family of receptor Serine/Threonine kinases. ACVR2A is involved in the activin and myostatin signaling pathway. (ACVR2A Aliases: ACVR2, ACTRII). These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling, and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Type II receptors are considered to be constitutively active kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR2A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ACVR2A
Gene Accession No. P27037, P27038, P38444
Gene Alias activin A receptor type 2A; activin A receptor type IIA; activin A receptor, type IIA; activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; Actrii; ActrIIa; ACTR-IIA; ACVR2; Acvr2a; rActR-II; TactrII; type II activin receptor
Gene Symbols ACVR2A
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACVR2A(Accession P27037), corresponding to amino acid residues V284-L334.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11480, 29263, 92
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.