Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACVR2A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA120174
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA120174 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA120174 Supplier Thermo Scientific Supplier No. PA120174

Goat Polyclonal Antibody

Recommended positive controls: muscle fibroblasts. Neutralization: when used at a 1-3 μg/mL, the antibody will block 50% of the binding of 30 ng/mL recombinant human activin A to immobilized recombinant human activin receptor IIA/Fc chimera (100 μL of a 0.5 μg/mL solution coating each well) in a functional ELISA assay.

ACVR2A Polyclonal antibody specifically detects ACVR2A in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR2A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and no preservative
Gene ACVR2A
Gene Accession No. P27037
Gene Alias activin A receptor type 2A; activin A receptor type IIA; activin A receptor, type IIA; activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; Actrii; ActrIIa; ACTR-IIA; ACVR2; Acvr2a; rActR-II; TactrII; type II activin receptor
Gene Symbols ACVR2A
Host Species Goat
Immunogen Recombinant fragment of human Activin Receptor Type IIA.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.