Learn More
Invitrogen™ ACVR2B Polyclonal Antibody

Description
Immunogen sequence: EGEQDKRLHC YASWRNSSGT IELVKKGCWL DDFNCYDRQE CVATEENPQV YFCCCEGNFC NERFTHLPEA GGPEVTYEPP PTAPTLLTVL AYSLLPIGGL S; Positive Samples: U-87MG, K-562; Cellular Location: Cell membrane, Single-pass type I membrane protein.
Specifications
Specifications
| Antigen | ACVR2B |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ACVR2B |
| Gene Accession No. | Q13705 |
| Gene Alias | activin A receptor type 2B; activin A receptor type IIB; activin A receptor, type IIB; activin receptor IIB; activin receptor type IIB; Activin receptor type-2B; Activine receptor 2b (transmembrane serine kinase); ActRIIB; ACTR-IIB; ActRIIB type II activin receptor B; ACVR2B; HTX4; MGC116908 |
| Gene Symbols | ACVR2B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.