Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVR2B Recombinant Rabbit Monoclonal Antibody (105)
GREENER_CHOICE

Catalog No. PIMA531028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531028 Supplier Invitrogen™ Supplier No. MA531028
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse ACVR2B.

ACVR2B is a type II member of the TGF-beta family of receptor ser/thr kinases and is involved in the activin and myostatin signaling pathway. Activins and inhibins are members of the TGF-beta superfamily due to amino acid homology with respect to the conservation of 7 of the 9 cysteine residues common to all TGF-beta forms. To date, seven type I and five type II activin receptors have been cloned from mammals, including activin receptor IA, activin receptor IIA, activin receptor IB, and activin receptor IIB. In addition, two splice variants of activin receptor IIA and five splice variants of activin receptor IIB have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVR2B
Applications ELISA
Classification Recombinant Monoclonal
Clone 105
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ACVR2B
Gene Accession No. P27040
Gene Alias activin A receptor type 2B; activin A receptor type IIB; activin A receptor, type IIB; activin receptor IIB; activin receptor type IIB; Activin receptor type-2B; Activine receptor 2b (transmembrane serine kinase); ActRIIB; ACTR-IIB; ActRIIB type II activin receptor B; ACVR2B; HTX4; MGC116908
Gene Symbols ACVR2B
Host Species Rabbit
Immunogen Recombinant Mouse ACVR2B Protein (Met1-Thr134).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11481
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.