Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVRL1 Monoclonal Antibody (05)
GREENER_CHOICE

Catalog No. PIMA530038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530038 Supplier Invitrogen™ Supplier No. MA530038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ALK-1/ACVRL1.

Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVRL1
Applications ELISA
Classification Monoclonal
Clone 05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ACVRL1
Gene Accession No. P37023
Gene Alias activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I
Gene Symbols ACVRL1
Host Species Mouse
Immunogen Recombinant Human ALK-1/ACVRL1 Protein (Met1-Gln118).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.