Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACVRL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118826
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118826 Supplier Invitrogen™ Supplier No. PA5118826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACVL1.

Activin A receptor type II-like 1 (ACVRL1) is a type I cell-surface receptor for the TGF-beta superfamily of ligands. ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2. Mutations in ACVRL1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACVRL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACVRL1
Gene Accession No. P37023, P80203, Q61288
Gene Alias activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I
Gene Symbols ACVRL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACVRL1(Accession P37023), corresponding to amino acid residues G211-A261.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11482, 25237, 94
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.