Learn More
ACY1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1B12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503227
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ACY1 encodes a cytosolic, homodimeric, zinc-binding enzyme that catalyzes the hydrolysis of acylated L-amino acids to L-amino acids and an acyl group, and has been postulated to function in the catabolism and salvage of acylated amino acids. This gene is located on chromosome 3p21.1, a region reduced to homozygosity in small-cell lung cancer (SCLC), and its expression has been reported to be reduced or undetectable in SCLC cell lines and tumors. The amino acid sequence of human aminoacylase-1 is highly homologous to the porcine counterpart, and this enzyme is the first member of a new family of zinc-binding enzymes. Mutations in this gene cause aminoacylase-1 deficiency, a metabolic disorder characterized by central nervous system defects and increased urinary excretion of N-acetylated amino acids. Alternative splicing of this gene results in multiple transcript variants. Read-through transcription also exists between this gene and the upstream ABHD14A (abhydrolase domain containing 14A) gene. A related pseudogene has been identified on chromosome 18.Specifications
| ACY1 | |
| Monoclonal | |
| Unconjugated | |
| ACY1 | |
| ACY-1, ACY1D, HEL-S-5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 109652, 300981, 476602, 95 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q03154, Q6AYS7, Q99JW2 | |
| Acy1 | |
| Full length human recombit protein of human ACY1 produced in HEk293 cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.