Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACY3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32142225UL
Description
ACY3 Polyclonal antibody specifically detects ACY3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ACY3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
ACY-3aspartoacylase-2, Acylase III, Aminoacylase-3, aspartoacylase (aminocyclase) 3, EC 3.5.1.-, EC 3.5.1.15, HCBP1ASPA2, Hepatitis C virus core-binding protein 1, MGC9740 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ACY3 | |
25 μg | |
Lipid and Metabolism | |
91703 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
0.2um filtered solution in PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction