Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACY3 Antibody (OTI2D5), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272329PECY7
Description
ACY3 Monoclonal specifically detects ACY3 in Human, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
ACY3 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
ACY3 | |
Full length recombinant protein of human ACY3 (NP_542389) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Primate, Monkey | |
IgG2b |
Flow Cytometry | |
OTI2D5 | |
Flow Cytometry | |
ACY-3aspartoacylase-2, Acylase III, Aminoacylase-3, aspartoacylase (aminocyclase) 3, EC 3.5.1.-, EC 3.5.1.15, HCBP1ASPA2, Hepatitis C virus core-binding protein 1, MGC9740 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
91703.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction