Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACY3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558700 Supplier Invitrogen™ Supplier No. PA558700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NTTANMGTCL IAKSSHEVFA MHLCRHLQLQ YPELSCQVFL YQRSGEESYN LDSVAKNGLG LELG Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 56%.

ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACY3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Acy3
Gene Accession No. Q96HD9
Gene Alias 0610006H10Rik; AA3; AAIII; Acy3; ACY-3; Acylase III; aminoacylase 3; Aminoacylase III; Aminoacylase-3; Aspa2; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; aspartoacylase-2; aspartoacylase-3; AW107362; EGK_05921; HCBP1; HCV core-binding protein 1; hepatitis C virus core-binding protein 1; LOW QUALITY PROTEIN: N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); RGD1305394
Gene Symbols Acy3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HCBP1. Recombinant protein control fragment (Product #RP-97319).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91703
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.