Learn More
                                    Invitrogen™ ACY3 Polyclonal Antibody
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592341
Description
Immunogen sequence: MCSLPVPREP LRRVAVTGGT HGNEMSGVYL ARHWLHAPAE LQRASFSAVP VLANPAATSG CRRYVDHDLN RTFTSSFLNS RPTPDDPYEV TRARELNQLL GPKASGQAFD FVLDLHNTTA NMGTCLIAKS SHEVFAMHLC RHLQLQYPEL SCQVFLYQRS GEESYNLDSV AKNGLGLELG PQPQGVLRAD IFSRMRTLVA TVLDFIELFN QGTAFPAFEM EAYRPVGVVD FPRTEAGHLA GTVHPQLQDR DFQPLQPGAP IFQMFSGEDL LYEGESTVYP VFINEAAYYE KGVAFVQTEK FTFTVPAMPA LTPAPSPAS; Positive Samples: Mouse kidney, Mouse heart; Cellular Location: Apical cell membrane, Cytoplasm, Peripheral membrane protein.
ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate. 
            Specifications
| ACY3 | |
| Polyclonal | |
| Unconjugated | |
| Acy3 | |
| 0610006H10Rik; AA3; AAIII; Acy3; ACY-3; Acylase III; aminoacylase 3; Aminoacylase III; Aminoacylase-3; Aspa2; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; aspartoacylase-2; aspartoacylase-3; AW107362; EGK_05921; HCBP1; HCV core-binding protein 1; hepatitis C virus core-binding protein 1; LOW QUALITY PROTEIN: N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); RGD1305394 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 71670 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| ELISA, Western Blot | |
| 2.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91XE4 | |
| Acy3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-319 of human ACY3 (NP_5423891). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
