Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acylglycerol Kinase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987172
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7172 Supplier Bioss Supplier No. BS6276R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lipid kinase that can phosphorylate both monoacylglycerol and diacylglycerol to form lysophosphatidic acid (LPA) and phosphatidic acid (PA), respectively. Does not phosphorylate sphingosine. Overexpression increases the formation and secretion of LPA, resulting in transactivation of EGFR and activation of the downstream MAPK signaling pathway, leading to increased cell growth.

Specifications

Antigen Acylglycerol Kinase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AGK
Gene Accession No. Q53H12
Gene Alias 2610037M15Rik; 6720408I04Rik; acylglycerol kinase; Acylglycerol kinase, mitochondrial; AGK; AI465370; CATC5; CTRCT38; FLJ10842; hAGK; HsMuLK; MTDPS10; Mulk; Multiple substrate lipid kinase; multi-substrate lipid kinase; RGD1562046
Gene Symbols AGK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Acylglycerol Kinase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55750
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.