Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACYP1 Antibody (004), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290125DL594
Description
ACYP1 Monoclonal specifically detects ACYP1 in Human samples. It is validated for ELISA.Specifications
ACYP1 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
97 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
4 | |
ELISA | |
acylphosphatase 1, erythrocyte (common) type, Acylphosphatase, erythrocyte isozyme, Acylphosphatase, organ-common type isozyme, Acylphosphate phosphohydrolase 1, ACYPEacylphosphatase-1, EC 3.6.1.7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ACYP1 (P07311; Ala 2-Lys 99). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction