Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACYP1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10057091
Description
ACYP1 Polyclonal specifically detects ACYP1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
ACYP1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
acylphosphatase 1, erythrocyte (common) type, Acylphosphatase, erythrocyte isozyme, Acylphosphatase, organ-common type isozyme, Acylphosphate phosphohydrolase 1, ACYPEacylphosphatase-1, EC 3.6.1.7 | |
Goat | |
Affinity Purified | |
RUO | |
97 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:128000 | |
P07311 | |
ACYP1 | |
Peptide with sequence C-PKSHIDKANFNNEK corresponding to internal region according to NP_001098.1. | |
0.1 mg | |
Primary | |
This antibody is expected to recognize isoform a (NP_001098.1) only. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction