Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACYP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518639
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518639 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518639 Supplier Invitrogen™ Supplier No. PA518639
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

Acylphosphatase is a small cytosolic enzyme that catalyzes the hydrolysis of the carboxyl-phosphate bond of acylphosphates. Two isoenzymes have been isolated, called muscle acylphosphatase and erythrocyte acylphosphatase, on the basis of their tissue localization. This gene encodes the erythrocyte acylphosphatase isoenzyme. Alternatively spliced transcript variants that encode different proteins were identified through data analysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACYP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ACYP1
Gene Accession No. P07311
Gene Alias 1110039O14Rik; acylphosphatase 1; acylphosphatase 1, erythrocyte (common) type; acylphosphatase, erythrocyte isozyme; Acylphosphatase, organ-common type isozyme; acylphosphatase-1; Acylphosphate phosphohydrolase 1; Acyp1; ACYPE; AI325944
Gene Symbols ACYP1
Host Species Goat
Immunogen Synthetic peptide sequence (PKSHIDKANFNNEK) corresponding to the internal amino acids of ACYP1 (aa 72-86).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 97
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.