Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACYP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557089 Supplier Invitrogen™ Supplier No. PA557089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEGNTLISV DYEIFGKVQG VFFRKHTQAE GKKLGLVGWV QNTDRGTVQG QLQGPISKVR HMQEWLETRG SPKSHIDKAN FNNEKVILKL DYSDFQI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

Acylphosphatase is a small cytosolic enzyme that catalyzes the hydrolysis of the carboxyl-phosphate bond of acylphosphates. Two isoenzymes have been isolated, called muscle acylphosphatase and erythrocyte acylphosphatase, on the basis of their tissue localization. This gene encodes the erythrocyte acylphosphatase isoenzyme. Alternatively spliced transcript variants that encode different proteins were identified through data analysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACYP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACYP1
Gene Accession No. P07311
Gene Alias 1110039O14Rik; acylphosphatase 1; acylphosphatase 1, erythrocyte (common) type; acylphosphatase, erythrocyte isozyme; Acylphosphatase, organ-common type isozyme; acylphosphatase-1; Acylphosphate phosphohydrolase 1; Acyp1; ACYPE; AI325944
Gene Symbols ACYP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACYP1. Recombinant protein control fragment (Product #RP-89113).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 97
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.