Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADA2a Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ADA2a |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ADA2a Polyclonal specifically detects ADA2a in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ADA2a | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, mTOR Pathway | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
6871 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MDRLGPFSNDPSDKPPCRGCSSYLMEPYIKCAECGPPPFFLCLQCFTRGFEYKKHQSDHTYEIMTSD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
ADA2AFLJ12705, ADA2-like protein, hADA2, TADA2LADA2, transcriptional adapter 2-alpha, Transcriptional adapter 2-like, transcriptional adaptor 2 (ADA2 homolog, yeast)-like, transcriptional adaptor 2 alpha, transcriptional adaptor 2A | |
TADA2A | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title