Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569115 Supplier Thermo Scientific Supplier No. PA569115
Add to Cart
Add to Cart

ADAD2 Polyclonal Antibody

ADAD2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAD2
Gene Accession No. Q8NCV1
Gene Alias 4930403J07Rik; ADAD2; adenosine deaminase domain containing 2; adenosine deaminase domain-containing protein 2; RGD1560949; TENRL; Testis nuclear RNA-binding protein-like
Gene Symbols ADAD2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ADAD2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161931
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.