Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32126225UL
Description
ADAL Polyclonal antibody specifically detects ADAL in Human samples. It is validated for ImmunofluorescenceSpecifications
ADAL | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
adenosine deaminase-like, adenosine deaminase-like protein, DKFZp313B2137, EC 3.5.4, EC 3.5.4.-, FLJ44620 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: DILMVTKDVIKEFADDGVKYLELRSTPRRENATGMTKKTYVESILEGIKQSKQENLDIDVRYLIAVDRRGGPLVAKETVKLAE | |
25 μg | |
DNA replication Transcription Translation and Splicing | |
161823 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction