Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543731 Supplier Invitrogen™ Supplier No. PA543731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ILLDLLPDRI GHGTFLNSGE GGSLDLVDFV RQHRIPLELC LTSNVKSQTV Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 92%; Human: 100%; Mouse: 79%; Pig: 100%; Rabbit: 93%; Rat: 86%; Zebrafish: 100%.

ADAL is a putative nucleoside deaminase. It may catalyze the hydrolytic deamination of adenosine or some similar substrate and play a role in purine metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADAL
Gene Accession No. Q6DHV7
Gene Alias 4930578F03Rik; Adal; adenosine deaminase like; adenosine deaminase-like; adenosine deaminase-like protein; Adenosine deaminase-like protein-like protein; hypothetical protein LOC619263; zgc:109912
Gene Symbols ADAL
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ADAL (aa 221-270).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161823
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.