Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112990
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112990 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112990 Supplier Invitrogen™ Supplier No. PA5112990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADAL Polyclonal Antibody for Western Blot, ELISA

ADAL is a putative nucleoside deaminase. It may catalyze the hydrolytic deamination of adenosine or some similar substrate and play a role in purine metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ADAL
Gene Accession No. Q6DHV7, Q80SY6
Gene Alias 4930578F03Rik; Adal; adenosine deaminase like; adenosine deaminase-like; adenosine deaminase-like protein; Adenosine deaminase-like protein-like protein; hypothetical protein LOC619263; zgc:109912
Gene Symbols ADAL
Host Species Rabbit
Immunogen Recombinant Human Adenosine deaminase-like protein (1-267AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161823, 75894
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.