Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Adalimumab (Humira™) (Human) ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
- Adalimumab (Humira™) (Human) ELISA Kit is a sandwich enzyme-linked immunoassay for the quantitative measurement of Adalimumab in human serum and plasma.
Specifications
Specifications
Cross Reactivity | Human |
Product Type | Kit |
Quantity | 1 Kit |
Storage Requirements | Store at 4°C |
Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction