Learn More
Adalimumab Mouse, Clone: 15C7, GenScript™
Mouse Monoclonal Antibody
Supplier: Genscript Corporation A0195440

Description
Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. Adalimumab, sold under the trade name Humira among others, is a medication used to treat rheumatoid arthritis, psoriatic arthritis, ankylosing spondylitis, Crohn's disease, ulcerative colitis, chronic psoriasis, hidradenitis suppurativa, and juvenile idiopathic arthritis. Adalimumab is a TNF-inhibiting, anti-inflammatory, biologic medication. It binds to tumor necrosis factor-alpha (TNFalpha), which normally binds to TNFalpha receptors, leading to the inflammatory response of autoimmune diseases. By binding to TNFalpha, adalimumab reduces this inflammatory response. Because TNFalpha is also part of the immune system, which protects the body from infection, treatment with adalimumab may increase the risk of infections.
Adalimumab Monoclonal antibody specifically detects Adalimumab in Chemical samples. It is validated for ELISASpecifications
Adalimumab | |
Monoclonal | |
Unconjugated | |
Humira | |
Adalimumab | |
40 μg | |
Primary | |
Antibody | |
IgG2b κ |
ELISA | |
15C7 | |
PBS with 0.02% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Chemical |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.