Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM10 Antibody (RM0146-7H12) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | ADAM10 |
---|---|
Clone | RM0146-7H12 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ADAM10 Monoclonal specifically detects ADAM10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
ADAM10 | |
LYOPH | |
Unconjugated | |
Rat | |
Apoptosis, Signal Transduction | |
O35598 | |
102 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0146-7H12 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian protein homolog, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
ADAM10 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title