Learn More
Invitrogen™ ADAM11 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ADAM11 protein.
Specifications
Specifications
| Antigen | ADAM11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ADAM11 |
| Gene Accession No. | O75078, Q9R1V4 |
| Gene Alias | a disintegrin and metallopeptidase domain 11; a disintegrin and metalloprotease domain (ADAM) 11; a disintegrin and metalloprotease domain 11; a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAM metallopeptidase domain 11; Adam11; ADAMs; AW060611; disintegrin and metalloproteinase domain-containing protein 11; Mdc; metalloendopeptidases; metalloproteinase-like, disintegrin-like, and cysteine-rich protein |
| Gene Symbols | ADAM11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.