Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAM11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584955
Description
Immunogen sequence: LPHLIYRTPL LPDPLGCREP GCLFAVPAQS APPNRPRLRR KRQVRRGHPT VHSETKYVEL IVINDHQLFE QMR.
Probable ligand for integrin in the brain. This is a non catalytic metalloprotease-like protein.
Specifications
ADAM11 | |
Polyclonal | |
Unconjugated | |
ADAM11 | |
a disintegrin and metallopeptidase domain 11; a disintegrin and metalloprotease domain (ADAM) 11; a disintegrin and metalloprotease domain 11; a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAM metallopeptidase domain 11; Adam11; ADAMs; AW060611; disintegrin and metalloproteinase domain-containing protein 11; Mdc; metalloendopeptidases; metalloproteinase-like, disintegrin-like, and cysteine-rich protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4185 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75078 | |
ADAM11 | |
Recombinant protein corresponding to Human ADAM11. Recombinant protein control fragment (Product #RP-108268). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction