Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986853
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6853 Supplier Bioss Supplier No. BS5846R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probable ligand for integrin in the brain. This is a non catalytic metalloprotease-like protein.

Specifications

Antigen ADAM11
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADAM11
Gene Accession No. O75078
Gene Alias a disintegrin and metallopeptidase domain 11; a disintegrin and metalloprotease domain (ADAM) 11; a disintegrin and metalloprotease domain 11; a disintegrin and metalloproteinase; ADAM; ADAM 11; ADAM metallopeptidase domain 11; ADAM11; ADAMs; AW060611; Disintegrin and metalloproteinase domain-containing protein 11; MDC; metalloendopeptidases; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein
Gene Symbols ADAM11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ADAM11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4185
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.