Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAM12 Monoclonal Antibody (3G1B10B9)
GREENER_CHOICE

Catalog No. PIMA530043
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530043 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530043 Supplier Thermo Scientific Supplier No. MA530043
Add to Cart
Add to Cart

ADAM12 Monoclonal Antibody (3G1B10B9)

ADAM12 Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM12
Applications ELISA
Classification Monoclonal
Clone 3G1B10B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ADAM12
Gene Accession No. O43184
Gene Alias a disintegrin and metallopeptidase domain 12 (meltrin alpha); a disintegrin and metalloprotease domain 12; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 12 (meltrin alpha); ADAM; ADAM 12; ADAM metallopeptidase domain 12; Adam12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin alpha; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; mKIAA4001; MLTN; Mltna; PRO545; RP11-295J3.5; UNQ346; UNQ346/PRO545
Gene Symbols ADAM12
Host Species Mouse
Immunogen Recombinant Human ADAM12 protein (Met1-Ser513).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8038
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.