Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583163 Supplier Invitrogen™ Supplier No. PA583163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLFTNKKTTI EKLRCVRPSR PPRGFQPCQA HLGHLGKGLM RKPPDSYPPK DNPRRLLQCQ NVDISRPLN Highest antigen sequence indentity to the following orthologs: Mouse - 61%, Rat - 61%.

ADAM12 is a member of the ADAM (a disintegrin and metalloprotease-like domain) family. Two forms of ADAM12 have been described: ADAM12S and ADAM12L (short and long forms). The short form is a soluble form lacking the transmembrane and cytoplasmic domains. The short form of ADAM12 was reported to provoke myogenesis, and the lack of cytoplasmic domain suggests different regulation pathways (although both forms can be expressed in the same tissue).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADAM12
Gene Accession No. O43184
Gene Alias a disintegrin and metallopeptidase domain 12 (meltrin alpha); a disintegrin and metalloprotease domain 12; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 12 (meltrin alpha); ADAM; ADAM 12; ADAM metallopeptidase domain 12; Adam12; ADAM12-OT1; ADAMs; CAR10; Disintegrin and metalloproteinase domain-containing protein 12; MCMP; MCMPMltna; meltrin alpha; meltrin-alpha; metalloendopeptidases; metalloprotease-disintegrin 12 transmembrane; mKIAA4001; MLTN; Mltna; PRO545; RP11-295J3.5; UNQ346; UNQ346/PRO545
Gene Symbols ADAM12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAM12. Recombinant protein control fragment (Product #RP-94992).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8038
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.