Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM15 Antibody (001), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290765APC
Description
ADAM15 Monoclonal antibody specifically detects ADAM15 in Mouse samples. It is validated for ELISASpecifications
ADAM15 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
001 | |
ELISA | |
ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ADAM15 (Accession#: O88839-1; Met1-Thr697). | |
0.1 mL | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
8751 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction