Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM15 Antibody (SR2269), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32222750UL
Description
ADAM15 Monoclonal antibody specifically detects ADAM15 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ADAM15 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR2269 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin | |
A synthesized peptide derived from human ADAM15 (Uniprot #: Q13444) | |
50 μg | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
8751 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction