Learn More
Invitrogen™ ADAM15 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547966
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse (rm) ADAM15 is observed, and less than 1% cross-reactivity with rmADAM9, rmADAM10, recombinant human (rh) ADAM8, rhADAM9, rhADAM10, rhADAM12, rhADAM17 (TACE), rhADAM19, rhADAM22, rhADAM23, rhADAM32, and rhADAM33 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the ADAM (a disintegrin and metalloproteinase) protein family. ADAM family members are type I transmembrane glycoproteins known to be involved in cell adhesion and proteolytic ectodomain processing of cytokines and adhesion molecules. This protein contains multiple functional domains including a zinc-binding metalloprotease domain, a disintegrin-like domain, as well as a EGF-like domain. Through its disintegrin-like domain, this protein specifically interacts with the integrin beta chain, beta 3. It also interacts with Src family protein-tyrosine kinases in a phosphorylation-dependent manner, suggesting that this protein may function in cell-cell adhesion as well as in cellular signaling. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| ADAM15 | |
| Polyclonal | |
| Unconjugated | |
| Adam15 | |
| a disintegrin and metallopeptidase domain 15 (metargidin); a disintegrin and metalloprotease domain 15 (metargidin); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 15; a disintegrin and metalloproteinase domain 15 (metargidin); AD56; ADAM; ADAM 15; ADAM metallopeptidase domain 15; Adam15; ADAMs; CRII-7; disintegrin and metalloprotease 15; disintegrin and metalloproteinase domain-containing protein 15; Mdc15; MDC-15; metalloendopeptidases; Metalloprotease RGD disintegrin protein; metalloproteinase-like, disintegrin-like, and cysteine-rich protein 15; metargidin; tMDC VI; tMDCVI | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8751 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q13444 | |
| Adam15 | |
| Sf 21-derived recombinant human ADAM15 Ectodomain Leu18-Ser693. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.