Learn More
Invitrogen™ ADAM15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596791
Description
Positive Samples: Mouse heart, Mouse kidney, Mouse brain; Cellular Location: Cell junction, Cell projection, Cytoplasmic vesicle, Endomembrane system, Single-pass type I membrane protein, acrosome, adherens junction, cilium, flagellum, secretory vesicle Immunogen sequence: QPAAPLCLQT ANTRGNAFGS CGRNPSGSYV SCTPRDAICG QLQCQTGRTQ PLLGSIRDLL WETIDVNGTE LNCSWVHLDL GSDVAQPLLT LPGTACGPGL VCIDHRCQRV DLLGAQECRS KCHGHGVCDS NRHCYCEEGW APPDCTTQLK ATSSLTTGLL L.
The protein encoded by this gene is a member of the ADAM (a disintegrin and metalloproteinase) protein family. ADAM family members are type I transmembrane glycoproteins known to be involved in cell adhesion and proteolytic ectodomain processing of cytokines and adhesion molecules. This protein contains multiple functional domains including a zinc-binding metalloprotease domain, a disintegrin-like domain, as well as a EGF-like domain. Through its disintegrin-like domain, this protein specifically interacts with the integrin beta chain, beta 3. It also interacts with Src family protein-tyrosine kinases in a phosphorylation-dependent manner, suggesting that this protein may function in cell-cell adhesion as well as in cellular signaling. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
ADAM15 | |
Polyclonal | |
Unconjugated | |
Adam15 | |
a disintegrin and metallopeptidase domain 15 (metargidin); a disintegrin and metalloprotease domain 15 (metargidin); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 15; a disintegrin and metalloproteinase domain 15 (metargidin); AD56; ADAM; ADAM 15; ADAM metallopeptidase domain 15; Adam15; ADAMs; CRII-7; disintegrin and metalloprotease 15; disintegrin and metalloproteinase domain-containing protein 15; Mdc15; MDC-15; metalloendopeptidases; Metalloprotease RGD disintegrin protein; metalloproteinase-like, disintegrin-like, and cysteine-rich protein 15; metargidin; tMDC VI; tMDCVI | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11490, 57025, 8751 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88839, Q13444, Q9QYV0 | |
Adam15 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 540-700 of human ADAM15 (NP_0038063). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.