Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM15 Recombinant Rabbit Monoclonal Antibody (007), APC
GREENER_CHOICE

Catalog No. PIMA540911
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540911 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540911 Supplier Invitrogen™ Supplier No. MA540911
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

The protein encoded by this gene is a member of the ADAM (a disintegrin and metalloproteinase) protein family. ADAM family members are type I transmembrane glycoproteins known to be involved in cell adhesion and proteolytic ectodomain processing of cytokines and adhesion molecules. This protein contains multiple functional domains including a zinc-binding metalloprotease domain, a disintegrin-like domain, as well as a EGF-like domain. Through its disintegrin-like domain, this protein specifically interacts with the integrin beta chain, beta 3. It also interacts with Src family protein-tyrosine kinases in a phosphorylation-dependent manner, suggesting that this protein may function in cell-cell adhesion as well as in cellular signaling. Multiple alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM15
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 007
Concentration 0.1 mg/mL
Conjugate APC
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene Adam15
Gene Accession No. Q13444
Gene Alias a disintegrin and metallopeptidase domain 15 (metargidin); a disintegrin and metalloprotease domain 15 (metargidin); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 15; a disintegrin and metalloproteinase domain 15 (metargidin); AD56; ADAM; ADAM 15; ADAM metallopeptidase domain 15; Adam15; ADAMs; CRII-7; disintegrin and metalloprotease 15; disintegrin and metalloproteinase domain-containing protein 15; Mdc15; MDC-15; metalloendopeptidases; Metalloprotease RGD disintegrin protein; metalloproteinase-like, disintegrin-like, and cysteine-rich protein 15; metargidin; tMDC VI; tMDCVI
Gene Symbols Adam15
Host Species Rabbit
Immunogen Recombinant Human ADAM15 protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8751
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.