Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA131074
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA131074 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA131074 Supplier Invitrogen™ Supplier No. PA131074
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX13535-PEP).. Expected to cross-react with Chinese Hamster due to sequence homology. Detects a band of approximately 125-130 kDa (predicted molecular weight: 95 or when glycosylated 130 kDa).

ADAM17 (disintegrin and metalloproteinase domain-containing protein 17) cleaves the membrane bound precusor of TNF-alpha to its mature soluble form. ADAM 17 is responsbile for the proteolytical release of soluble JAM3 from endothelial cells surface and the proteolytic release of several other cell-surface proteins, including p75 TNF-receptor, interleukin receptor type II, p55 TNF-receptor, transforming growth factor-alpha, L-selectin, growth hormone receptor, MUC1, and the amyloid precursor protein. It acts as an activator of the Notch pathway by mediating cleavage of Notch and generating the membrane-associated intermediate fragment called Notch extracellular truncation (NEXT). Mutations in the gene can result in inflammatory skin and bowel disease, neonatal 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ADAM17
Gene Accession No. P78536
Gene Alias a disintegrin and metallopeptidase domain 17; a disintegrin and metalloprotease domain 17; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 17; a disintegrin and metalloproteinase domain 17 (tumor necrosis factor, alpha, converting enzyme); ADAM; ADAM 17; ADAM metallopeptidase domain 17; ADAM metallopeptidase domain 18; Adam17; ADAM18; ADAMs; CD156B; CD156b antigen; CSVP; Disintegrin and metalloproteinase domain-containing protein 17; metalloendopeptidases; NISBD; NISBD1; Snake venom-like protease; TACE; TNF-alpha convertase; TNF-alpha converting enzyme; TNF-alpha-converting enzyme; tumor necrosis factor, alpha, converting enzyme
Gene Symbols ADAM17
Host Species Goat
Immunogen Peptide with sequence LQRQNRVDSKETEC, from the C Terminus.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6868
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.