Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169367
Description
ADAM19 Polyclonal specifically detects ADAM19 in Human samples. It is validated for Western Blot.Specifications
ADAM19 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
a disintegrin and metalloproteinase domain 19 (meltrin beta), ADAM 19, ADAM metallopeptidase domain 19, disintegrin and metalloproteinase domain-containing protein 19, EC 3.4.24, EC 3.4.24.-, MADDAM, Meltrin-beta, Metalloprotease and disintegrin dendritic antigen marker, MLTNBmeltrin beta | |
Rabbit | |
82 kDa | |
100 μL | |
Dendritic Cell Markers | |
8728 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9H013 | |
ADAM19 | |
Synthetic peptides corresponding to ADAM19(ADAM metallopeptidase domain 19 (meltrin beta)) The peptide sequence was selected from the N terminal of ADAM19. Peptide sequence GRELILDLEKNEQLFAPSYTETHYTSSGNPQTTTRKLEDHCFYHGTVRET. | |
Affinity purified | |
RUO | |
Primary | |
Canine: 86%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction