Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143125
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143125 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143125 Supplier Invitrogen™ Supplier No. PA5143125
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a type I transmembrane protein and serves as a marker for dendritic cell differentiation. It has also been demonstrated to be an active metalloproteinase, which may be involved in normal physiological and pathological processes such as cells migration, cell adhesion, cell-cell and cell-matrix interactions, and signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ADAM19
Gene Accession No. Q9H013
Gene Alias a disintegrin and metallopeptidase domain 19 (meltrin beta); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 19 (meltrin beta); ADAM; ADAM 19; ADAM metallopeptidase domain 19; ADAM19; ADAMs; AL024287; disintegrin and metalloproteinase domain-containing protein 19; FKSG34; MADDAM; meltrin beta; meltrin-beta; metalloendopeptidases; Metalloprotease and disintegrin dendritic antigen marker; metalloprotease-disintegrin meltrin beta; MLTNB; Sox30; SRY-box containing gene 30
Gene Symbols ADAM19
Host Species Goat
Immunogen Peptide with sequence KIQCQSSEARPLESN.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8728
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.