Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAM19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA125175
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a type I transmembrane protein and serves as a marker for dendritic cell differentiation. It has also been demonstrated to be an active metalloproteinase, which may be involved in normal physiological and pathological processes such as cells migration, cell adhesion, cell-cell and cell-matrix interactions, and signal transduction.
Specifications
ADAM19 | |
Polyclonal | |
Unconjugated | |
ADAM19 | |
a disintegrin and metallopeptidase domain 19 (meltrin beta); a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 19 (meltrin beta); ADAM; ADAM 19; ADAM metallopeptidase domain 19; ADAM19; ADAMs; AL024287; disintegrin and metalloproteinase domain-containing protein 19; FKSG34; MADDAM; meltrin beta; meltrin-beta; metalloendopeptidases; Metalloprotease and disintegrin dendritic antigen marker; metalloprotease-disintegrin meltrin beta; MLTNB; Sox30; SRY-box containing gene 30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8728 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% sodium azide | |
Q9H013 | |
ADAM19 | |
Synthetic peptide corresponding to the amino end of the propeptide domain of human ADAM19. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction