Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ADAM2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ADAM2 Polyclonal specifically detects ADAM2 in Human samples. It is validated for Western Blot.Specifications
ADAM2 | |
Polyclonal | |
Rabbit | |
Human | |
ADAM metallopeptidase domain 2, Cancer/testis antigen 15, CRYN2, CT15ADAM 2, disintegrin and metalloproteinase domain-containing protein 2, EC 3.4.24, fertilin beta, Fertilin subunit beta, FTNBCRYN1, PH-30, PH-30b, PH30PH30-beta | |
ADAM2 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q99965 | |
2515 | |
Synthetic peptides corresponding to ADAM2(ADAM metallopeptidase domain 2 (fertilin beta)) The peptide sequence was selected from the middle region of ADAM2. Peptide sequence PHDVAFLLVYREKSNYVGATFQGKMCDANYAGGVVLHPRTISLESLAVIL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title