Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADAM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555126 Supplier Invitrogen™ Supplier No. PA555126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CENCLFMSKE RMCRPSFEEC DLPEYCNGSS ASCPENHYVQ TGHPCGLNQW ICIDGVCMSG DK Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 48%.

ADAM2 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM2 is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions. This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Adam2
Gene Accession No. Q99965
Gene Alias a disintegrin and metallopeptidase domain 2; a disintegrin and metalloprotease domain 2; a disintegrin and metalloprotease domain (ADAM) 2; a disintegrin and metalloprotease domain 2 (fertilin beta); a disintegrin and metalloproteinase; ADAM; ADAM 2; ADAM metallopeptidase domain 2; Adam2; ADAMs; AI323749; cancer/testis antigen 15; CRYN1; CRYN2; CT15; Disintegrin and metalloproteinase domain-containing protein 2; fertilin beta; fertilin subunit beta; FTNB; metalloendopeptidases; PH30; PH-30; PH-30b; Ph30-beta
Gene Symbols Adam2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADAM2. Recombinant protein control fragment (Product #RP-93765).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2515
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.