Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADAM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578725
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SMMC Cell.
ADAM2 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM2 is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions. This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This member is a subunit of an integral sperm membrane glycoprotein called fertilin, which plays an important role in sperm-egg interactions.
Specifications
ADAM2 | |
Polyclonal | |
Unconjugated | |
Adam2 | |
a disintegrin and metallopeptidase domain 2; a disintegrin and metalloprotease domain 2; a disintegrin and metalloprotease domain (ADAM) 2; a disintegrin and metalloprotease domain 2 (fertilin beta); a disintegrin and metalloproteinase; ADAM; ADAM 2; ADAM metallopeptidase domain 2; Adam2; ADAMs; AI323749; cancer/testis antigen 15; CRYN1; CRYN2; CT15; Disintegrin and metalloproteinase domain-containing protein 2; fertilin beta; fertilin subunit beta; FTNB; metalloendopeptidases; PH30; PH-30; PH-30b; Ph30-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2515 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q99965 | |
Adam2 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human ADAM2 (701-715aa VLIAIMVKVNFQRKK). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction